Natriumhydroxid, micropearls for analysis
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:318 °C
- Density:2,13 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
3 Options Available Below
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:318 °C
- Density:2,13 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
ACRO38304
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Pearls
- Titration with HCl:≥99.0 %
- Sodium carbonate (Na2CO3):≤0.4 %
- Sodium sulfate (Na2SO4):≤80 ppm
- Sodium chloride (NaCl):≤200 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤0.5 ppm