Orders LinkContactUsLinkComponent
Natriumazid
Natriumazid
Toxic
Environment
Supplier:   PanReac AppliChem
Natriumazid
Supplier:  PanReac AppliChem
Certificates
2 Options Available Below
Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Merck Index:13,08653
  • Melting Pt:275 °C
  • Density:1,85 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:247-852-1
  • UN:1687
  • ADR:6.1,II
Documents
Certificates
Search by Lot Number
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Plastic bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 720,00 kr
    720,00 kr
    1 * 100 g
       720,00 kr
  • Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 2.360,00 kr
    2.360,00 kr
    1 * 500 g
       2.360,00 kr