Orders LinkContactUsLinkComponent
Natriumiodid, Ultrapure
Natriumiodid, Ultrapure
Environment
Supplier:   VWR Chemicals
Natriumiodid, Ultrapure
Supplier:  VWR Chemicals
Certificates
Natriumiodid, Ultrapure
Natriumiodid, Ultrapure
2 Options Available Below
Product Details & Documents
Synonyms:  Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
  • CAS Number:7681-82-5
  • Formula:NaI
  • MDL Number:MFCD00003532
  • Molecular Weight:149.89 g/mol
  • Merck Index:13,08705
  • Boiling Pt:1300 °C (1013 hPa)
  • Melting Pt:662 °C
  • Density:3,67 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:231-679-3
  • UN:3077
  • ADR:9,III
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • HDPE bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 2.420,00 kr
    2.420,00 kr
    1 * 250 g
       2.420,00 kr
  • HDPE bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 3.360,00 kr
    3.360,00 kr
    1 * 500 g
       3.360,00 kr