Natriumchlorid
Glass bottle for solids, 1 kg
SERA30183.01
Marketsource
: Serva
: Saline, NaCl, Sea salt, Halite
- :7647-14-5
- :NaCl
- :MFCD00003477
- :58.44 g/mol
- :14,08599
- :1413 °C (1013 hPa)
- :801 °C
- :2,16 g/cm³ (20 °C)
- :Stue temperatur
- :231-598-3
- Pakning:Glass bottle for solids
- Pakn:1 kg