Table clamp for rods
Hærdet støbejern, 12/13 mm, Stave
Katalog # BOCH5260
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Large table clamp for rods, made of tempered cast iron.
- Can be attached to table tops with a thickness of max. 55 mm
- Green varnished
Specifikationer
- Til:Stave
- Spændvidde:12/13 mm
- Materiale:Hærdet støbejern