Kaliumchlorid
Glass bottle for solids, 1 kg
Katalog # SERA26868.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Potassiumchloride
- CAS nummer:7447-40-7
- Formel:KCl
- MDL nummer:MFCD00011360
- Molecular Weight:74.55 g/mol
- Merck Index:14,07621
- Kogende Pt:1420 °C (1013 hPa)
- Smeltende Pt:773 °C
- Tæthed:1,98 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-211-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg