Natriumcarbonat
Glass bottle for solids, 1 kg
Katalog # SERA30181.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Calcined soda, Carbonic acid disodium salt
- CAS nummer:497-19-8
- Formel:Na₂CO₃
- MDL nummer:MFCD00003494
- Molecular Weight:105.99 g/mol
- Merck Index:14,08596
- Kogende Pt:1600 °C (1013 hPa)
- Smeltende Pt:854 °C
- Tæthed:2,53 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:207-838-8
- NÅ:01-2119485498-19
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg