Natriumcarbonat, Supelco®
Glass bottle for solids, 50 g
Katalog # SUPL71363-50G
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Calcined soda, Carbonic acid disodium salt
- CAS nummer:497-19-8
- Formel:Na₂CO₃
- MDL nummer:MFCD00003494
- Molecular Weight:105.99 g/mol
- Merck Index:14,08596
- Kogende Pt:1600 °C (1013 hPa)
- Smeltende Pt:854 °C
- Tæthed:2,53 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:207-838-8
- NÅ:01-2119485498-19
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:50 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



