Natriumborhydrid
Glass bottle for solids, 25 g
Katalog # SERA30182.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Sodium tetrahydridoborate, Sodium tetrahydroborate
- CAS nummer:16940-66-2
- Formel:NaBH₄
- MDL nummer:MFCD00003518
- Molecular Weight:37.83 g/mol
- Merck Index:13,08664
- Smeltende Pt:400 °C
- Tæthed:1,07 g/cm³ (20 °C)
- Flash Pt:69 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:241-004-4
- FN:1426
- ADR:4.3,I
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



