(R)-(+)-α-Terpineol, Supelco®
Glass bottle, 5 ml
Katalog # 83073-5ML-F
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: (R)-2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
- CAS nummer:7785-53-7
- Formel:C₁₀H₁₈O
- MDL nummer:MFCD00171435
- Molecular Weight:154.25 g/mol
- :M-Clarity™ quality level = MQ100
- EINECS:232-081-5
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:5 ml



