Pesticid referencestandard, Tributyltinchlorid 100 µg/ml i dichlormethan
Katalog # CAMBULM-8061-1.2
Leverandør: CAMBRIDGE ISOTOPE LABS
CAS nummer:
Specifikationer
- Pakning:Glass bottle
- Pakn:1,2 mL
- CAS-nr:1461-22-9
- Beskrivelse:Tributyltinchlorid 100 µg/ml i dichlormethan
- Formel:C₁₂H₂₇ClSn
- Beskrivelse:Tributyltin chloride