Organisk referensestandard, 1,8-Dimethylnaphthalen 50 µg/ml i toluen
CAMBULM-6181-1.2
:
- Pakning:Glass bottle
- Pakn:1,2 mL
- CAS-nr:569-41-5
- Beskrivelse:1,8-Dimethylnaphthalen 50 µg/ml i toluen
- Formel:C₁₂H₁₂
- Beskrivelse:1,8-Dimethylnaphthalene