Orders LinkContactUsLinkComponent
Stirrer bearings, KPG®
Stirrer bearings, KPG®
Stirrer bearing, HB 16, DURAN®, Shaft - Ø: 16 mm
Katalog # LENZ06300116
Leverandør:   Lenz Laborglas
Stirrer bearings, KPG®
Katalog # LENZ06300116
Leverandør:  Lenz Laborglas
Stirrer bearings, KPG®
Stirrer bearings, KPG®
Stirrer bearing, HB 16, DURAN®, Shaft - Ø: 16 mm
Price
542,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These stirrer bearings are made of precision calibrated glass tube.
  • Surface is grounded and polished
  • Interchangeable
Documents
Specifikationer
  • Til:
    Shaft - Ø: 16 mm
  • Beskrivelse:
    Stirrer bearing, HB 16, DURAN®
  • Seq:
    03