L(+)-Citrullin
Glass bottle for solids, 100 g
Katalog # SERA17311.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: H-Cit-OH
- CAS nummer:372-75-8
- Formel:C₆H₁₃N₃O₃
- MDL nummer:MFCD00064397
- Molecular Weight:175.19 g/mol
- EINECS:206-759-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



