Metanil gul
Glass bottle for solids, 25 g
Katalog # CHRO1B-321
Marketsource
Leverandør: CHROMA
Gå til:
Product Details & Documents
Synonymer: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
- CAS nummer:587-98-4
- Formel:C₁₈H₁₄N₃NaO₃S
- MDL nummer:MFCD00007486
- Molecular Weight:375.38 g/mol
- EINECS:209-608-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



