Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Metanil gul

Metanil gul

Glass bottle for solids, 25 g
Katalog # CHRO1B-317
Marketsource
Leverandør:   CHROMA
Metanil gul
Katalog # CHRO1B-317
Leverandør:  CHROMA
Marketsource
CAS nummer:  587-98-4
Metanil gul

Glass bottle for solids, 25 g

CAS nummer:  587-98-4
Price
Spørg for pris
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
  • CAS nummer:587-98-4
  • Formel:C₁₈H₁₄N₃NaO₃S
  • MDL nummer:MFCD00007486
  • Molecular Weight:375.38 g/mol
  • EINECS:209-608-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.