Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type, DURAN®
Sedimentation cones, Imhoff type, DURAN®
Imhoff-kolber, 1000 ml, uden stophane
Katalog # SCOT214005402
Leverandør:   DWK Life Sciences
Sedimentation cones, Imhoff type, DURAN®
Katalog # SCOT214005402
Leverandør:  DWK Life Sciences
Sedimentation cones, Imhoff type, DURAN®
Sedimentation cones, Imhoff type, DURAN®
Imhoff-kolber, 1000 ml, uden stophane
Price
8.390,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Without graduations
  • Very good chemical resistance
  • Very good temperature resistance
Specifikationer
  • Model:
    uden stophane
  • Beskrivelse:
    Imhoff-kolber, 1000 ml