Orders LinkContactUsLinkComponent
Equipment table
Equipment table
Bord til instrumenter + udstyr, mobilt med 4 drejelige hjul, antistatiske, 625×425×800 mm
Katalog # HUPF7500204
Leverandør:   HUPFER METALLWERKE
Equipment table
Katalog # HUPF7500204
Leverandør:  HUPFER METALLWERKE
Equipment table
Equipment table
Bord til instrumenter + udstyr, mobilt med 4 drejelige hjul, antistatiske, 625×425×800 mm
Price
8.970,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The instrument table is made from stainless steel. It is a solid, jointless tube structure made from square-type tube (20×20 mm), with two removable table tops formed as protection gallery (border top).
  • Electro-conductive
  • With double ball bearing the thread protection
  • Castors with 75 mm diameter
The instrument table is to provide immersion trays, sterile goods containers and baskets in operating theatres and neighbouring areas.
Certifications: Conforms with DIN 90952 packaging for sterilising goods.
Specifikationer
  • Beskrivelse:
    Bord til instrumenter + udstyr, mobilt med 4 drejelige hjul, antistatiske
  • B×D×H:
    625×425×800 mm