Soap dispenser
Sæbedispenser, 950 ml, 76×120×298 mm
Katalog # HUPF8600025
Leverandør: HUPFER METALLWERKE
Gå til:
Product Details & Documents
Soap dispenser made from stainless steel for exchangeable bottles in food production or logistics subject to rigorous levels of hygiene.
- Refillable tank
- Lockable filling cap
- Consistent cleanliness in all areas of operation
For use in kitchens to ensure uncompromised safety.
Specifikationer
- Kapacitet:950 ml
- Beskrivelse:Sæbedispenser
- B×D×H:76×120×298 mm