Orders LinkContactUsLinkComponent
Pharmacy scoops
Pharmacy scoops
Supplier:   BOCHEM
Pharmacy scoops
Supplier:  BOCHEM
Certificates
Pharmacy scoops
2 Options Available Below
Product Details & Documents
Stainless steel 18/10.
  • Soldered handle
Product Family Options
Product Information
  • Kapacitet
    Længde
    Ø
    Availability
    Price
  • 100 ml
    220 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 957,00 kr
    957,00 kr
    1 * 1 Items
       957,00 kr
  • 400 ml
    280 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.060,00 kr
    1.060,00 kr
    1 * 1 Items
       1.060,00 kr