Orders LinkContactUsLinkComponent
Cryostorage boxes, Cryostore™
Cryostorage boxes, Cryostore™
Supplier:   Simport Scientific
Cryostorage boxes, Cryostore™
Supplier:  Simport Scientific
Certificates
Cryostorage boxes, Cryostore™
Cryostorage boxes, Cryostore™
7 Options Available Below
Product Details & Documents
PC, coloured.
  • Writable surfaces
  • Available with four colours
  • Printed with squared numbers
  • Transparent cover allows to see the content of the box
These durable cryogenic storage boxes are designed to be used at temperatures between −196 °C and +121 °C. It can accommodate around 42×T310-10A Cryovials®.
Documents
Product Family Options
Product Information
  • Rum
    Til
    Farve
    B×D×H udv.
    Availability
    Price
  • 5×5
    1 - 2 ml cryotubes
    blå
    76×76×52 mm
    Log in for availability
    Log in for availability
    1 * 8 Items
    1 * 8 Items - kr562.00
    kr562.00
    1 * 8 Items
       kr562.00
  • 9×9
    3 - 4 ml cryotubes
    blå
    133×133×81 mm
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - kr1,120.00
    kr1,120.00
    1 * 6 Items
       kr1,120.00
  • 7×6
    Kryorør, 10 ml
    blå
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - kr1,560.00
    kr1,560.00
    1 * 10 Items
       kr1,560.00
  • 7×6
    10 ml cryotubes
    grøn
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - kr1,560.00
    kr1,560.00
    1 * 10 Items
       kr1,560.00
  • 7×6
    Kryorør, 10 ml
    rød
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - kr1,560.00
    kr1,560.00
    1 * 10 Items
       kr1,560.00
  • 7×6
    Kryorør, 10 ml
    gul
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - kr1,560.00
    kr1,560.00
    1 * 10 Items
       kr1,560.00
  • 9×9
    3 - 5 ml cryotubes
    blå
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - kr929.00
    kr929.00
    1 * 5 Items
       kr929.00
  • Discontinued