Removable needles
Om denne vare
The benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line. The result is a zero dead volume connection that simplifies priming.
Custom lengths and point styles are available. You can order your custom made needle according to your specification.
* Large hub
** Kel-F hub
83 tilgængelige muligheder nedenfor
Product Details & Documents
The benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line. The result is a zero dead volume connection that simplifies priming.
Custom lengths and point styles are available. You can order your custom made needle according to your specification.
* Large hub
** Kel-F hub
Documents
Product Family Options
RN fused silica needles for 5 - 10 µl syringes
Produktinformation
RN PEEK needles for 250 µl - 10 ml syringes
Produktinformation
RN needles for 2,5 - 100 µl syringes
Produktinformation
- MålerLængdeUdv.ØØ indv.SpidstypeTilgængelighedPrice
Custom made needles for 2,5 - 100 µl syringes
Produktinformation
- MålerLængdeUdv.ØØ indv.SpidstypeTilgængelighedPrice
- Specifikationer:New ProductNew Product
Custom made needles for 250 µl - 10 ml syringes
Produktinformation
- MålerLængdeUdv.ØØ indv.SpidstypeTilgængelighedPrice
RN needles for 250 µl - 10 ml syringes
Produktinformation
- MålerLængdeUdv.ØØ indv.SpidstypeTilgængelighedPrice
Small hub RN CTC needles
Produktinformation
- MålerLængdeUdv.ØØ indv.SpidstypeTilgængelighedPrice