Vacuum hand pump
Vacuum pump with manometer, Plastic tubing
Katalog # SART16673
Leverandør: Sartorius
Gå til:
Product Details & Documents
The hand operated vacuum pump made of PVC is used as practical vacuum source for outside of a laboratory.
Leveringsinformation: Supplied completely with gauge, vacuum release lever and a clear plastic tubing (600 mm).
Specifikationer
- Beskrivelse:Vacuum pump with manometer
- Kabinet:Plastic tubing