Orders LinkContactUsLinkComponent
Methylboronsyre
Methylboronsyre
Glass bottle for solids, 25 g
Katalog # COBBBB-3469-25G
Marketsource
Leverandør:   COMBI-BLOCKS
Methylboronsyre
Katalog # COBBBB-3469-25G
Leverandør:  COMBI-BLOCKS
Marketsource
CAS nummer:  13061-96-6
Methylboronsyre
Glass bottle for solids, 25 g
CAS nummer:  13061-96-6
Price
126,42 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
  • CAS nummer:13061-96-6
  • Formel:CH₅BO₂
  • MDL nummer:MFCD00002105
  • Molecular Weight:59.86 g/mol
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g