Dissektionssæt
Dissektionssæt, seks dele
631-1279
: RSG
Dissecting set with six parts made from stainless steel, packed in a pouch.
- 1 spoon spatula, 150 mm
- 1 pointed tweezer, 130 mm
- 1 powder spatula, 150/9 mm
- 1 scalpel, 125 mm
- 1 scissors straight, 100 mm
- 1 spatula with blade, 150 mm
- Beskrivelse:Dissektionssæt, seks dele