Beaker tongs, stainless steel, polished
230 mm, 15 - 60 mm
Katalog # RSGA043.230
Leverandør: RSG
Gå til:
Product Details & Documents
These beaker tongs are made from electrolytically polished stainless steel.
- Jaws with cork inserts
Specifikationer
- Længde:230 mm
- Dækket område:15 - 60 mm