ClipTip™ Pipette Tips
Om denne vare
ClipTip™ pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
- Pipette tips feature 'clips' that lock tips in place and forms a complete seal
- Every ClipTip™ pipette tip utilises low retention technology to enable maximium sample recovery
- Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
- Each tip insert has ink-jetted lot and volume information; the convenient colour key makes it easy to match the colour coding of the F1-ClipTip™ and E1-ClipTip™ pipettes with the corresponding ClipTip™ pipette tip rack insert
- Environmentally friendly reload system uses recycled components and 30% less plastic
Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure.
ClipTip™ pipette tips feature 'clips' that lock tips in place and form a complete seal, Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure, ClipTip™ pipette tips are clipped securely on the F1-ClipTip™ and E1-ClipTip™ pipettes with a light touch eliminating the frustration of banging tips on your pipette, Low retention technology minimizes liquid retention, and enable maximium sample recovery.
40 tilgængelige muligheder nedenfor
Product Details & Documents
ClipTip™ pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
- Pipette tips feature 'clips' that lock tips in place and forms a complete seal
- Every ClipTip™ pipette tip utilises low retention technology to enable maximium sample recovery
- Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
- Each tip insert has ink-jetted lot and volume information; the convenient colour key makes it easy to match the colour coding of the F1-ClipTip™ and E1-ClipTip™ pipettes with the corresponding ClipTip™ pipette tip rack insert
- Environmentally friendly reload system uses recycled components and 30% less plastic
Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure.
ClipTip™ pipette tips feature 'clips' that lock tips in place and form a complete seal, Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure, ClipTip™ pipette tips are clipped securely on the F1-ClipTip™ and E1-ClipTip™ pipettes with a light touch eliminating the frustration of banging tips on your pipette, Low retention technology minimizes liquid retention, and enable maximium sample recovery.
Documents
Certifikater
Søg efter partinummerAppNote_KFDuoPrime_E1CT Eq_012016 AT-Smart Note E1-ClipTip Adjustable Tip Spacing_RevA AT-Thermo_Scientific_ClipTip_Technology_Transform Daily Pipetting-AppNote1 E1-ClipTip Tip Attachment and Ejection Forces Smart Note Thermo Scientific E1-ClipTip Gel Loading for PCR
Application Notes
BRHPCLIPEUVWR0916_V2
Product Literature
Specifications for Product Family
Specifikationer
- Catalog No:
- 613-6031
- 613-6032
- 613-6025
- 613-6026
- 613-6861
- 613-6862
- 613-6192
- 613-6193
- 613-2205
- 613-2206
- 613-2207
- 613-2208
- 613-6027
- 613-6028
- 613-2209
- 613-2210
- 613-2211
- 613-2212
- 613-6029
- 613-6030
- 613-2213
- 613-2214
- 613-2215
- 613-2216
- 613-2217
- 613-2218
- 613-2219
- 613-2220
- 613-6194
- 613-6195
- 613-7447
- 613-7446
- 613-2222
- 613-2223
- 613-2224
- 613-2225
- 613-6033
- 613-6034
- 613-6035
- 613-6036
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Miljøcertificering:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
Specifikationer
Product Family Options
ClipTip 12,5
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Standard0,1 - 12,5 µl-30 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 348,00 kr348,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items348,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6031Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Standard0,1 - 12,5 µl+30 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 451,00 kr451,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items451,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6032Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Standard0,1 - 12,5 µl-30 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.290,00 kr2.290,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.290,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6025Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Standard0,1 - 12,5 µl+30 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.700,00 kr2.700,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.700,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6026Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Extended0,1 - 12,5 µl-41 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 342,00 kr342,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items342,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6861Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Extended0,1 - 12,5 µl+41 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 443,00 kr443,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items443,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6862Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Lav fremstillingspåvirkning - Extended0,1 - 12,5 µl-41 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.240,00 kr2.240,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.240,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6192Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Extended0,1 - 12,5 µl+41 mmlyserødE1-ClipTip and F1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.640,00 kr2.640,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.640,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6193Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 20
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Low Retention1 - 20 µl-46 mmlyserødF1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 348,00 kr348,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items348,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2205Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention1 - 20 µl+46 mmlyserødF1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 451,00 kr451,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items451,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2206Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention1 - 20 µl-46 mmlyserødF1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 323,00 kr323,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items323,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2207Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention1 - 20 µl+46 mmlyserødF1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 415,00 kr415,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items415,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2208Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 30
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Standard 3841 - 30 µl-44 mmVioletE1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.290,00 kr2.290,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.290,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6027Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Standard 3841 - 30 µl+44 mmVioletE1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.700,00 kr2.700,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.700,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6028Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 50
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Low Retention5 - 50 µl-51 mmVioletF1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 348,00 kr348,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items348,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2209Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention5 - 50 µl+51 mmVioletF1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 448,00 kr448,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items448,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2210Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention5 - 50 µl-51 mmVioletF1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 323,00 kr323,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items323,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2211Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention5 - 50 µl+51 mmVioletF1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 415,00 kr415,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items415,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2212Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 125
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Standard 3841 - 125 µl-64 mmgulE1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.250,00 kr2.250,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.250,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6029Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Standard 3841 - 125 µl+64 mmgulE1-ClipTip pipettes10 racks, 384 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 3.840 Items1 * 3.840 Items - 2.650,00 kr2.650,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 3.840 Items2.650,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6030Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 200
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Low Retention10 - 200 µl-56 mmgulE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 346,00 kr346,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items346,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2213Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention10 - 200 µl+56 mmgulE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 448,00 kr448,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items448,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2214Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention10 - 200 µl-56 mmgulE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 323,00 kr323,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items323,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2215Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention10 - 200 µl+56 mmgulE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 415,00 kr415,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items415,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2216Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 300
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Low Retention30 - 300 µl-63 mmorangeE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 346,00 kr346,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items346,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2217Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention30 - 300 µl+63 mmorangeE1-ClipTip and F1-ClipTip pipettes10 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 448,00 kr448,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items448,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2218Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention30 - 300 µl-63 mmorangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 323,00 kr323,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items323,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2219Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention30 - 300 µl+63 mmorangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 415,00 kr415,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items415,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2220Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention, Extended Length30 - 300 µl-100 mmorangeE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog ind for tilgængelighed1 * 768 Items483,00 krSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6194Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention, Extended Length30 - 300 µl+100 mmorangeE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 370,00 kr370,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items370,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6195Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention, Extended Length30 - 300 µl+100 mmorangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 427,00 kr427,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items427,00 krSpecifikationer:More
Specifications
Cat. No.613-7447Environmentally PreferableMiljøcertificering - Low Retention, Extended Length30 - 300 µl-100 mmorangeE1-ClipTip and F1-ClipTip pipettes10 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 960 Items1 * 960 Items - 332,00 kr332,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 960 Items332,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-7446Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 1000
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Low Retention100 - 1000 µl-95 mmblåE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 381,00 kr381,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items381,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2222Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention100 - 1000 µl+95 mmblåE1-ClipTip and F1-ClipTip pipettes8 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 492,00 kr492,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items492,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2223Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention100 - 1000 µl-95 mmblåE1-ClipTip and F1-ClipTip pipettes8 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 357,00 kr357,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items357,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2224Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention100 - 1000 µl+95 mmblåE1-ClipTip and F1-ClipTip pipettes8 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 451,00 kr451,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items451,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-2225Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
ClipTip 1250
Produktinformation
- SpidstypeVolumenSterilLængdeFarvekodeTil brug medPakketAvailabilityPrice
- Low Retention15 - 1250 µl-103 mmturkisE1-ClipTip pipettes8 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 377,00 kr377,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items377,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6033Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention15 - 1250 µl+103 mmturkisE1-ClipTip pipettes8 racks, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 485,00 kr485,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items485,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6034Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention15 - 1250 µl-103 mmturkisE1-ClipTip pipettes8 reload inserts, 96 eachLog ind for tilgængelighed1 * 768 Items594,00 krSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6035Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning - Low Retention15 - 1250 µl+103 mmturkisE1-ClipTip pipettes8 reload inserts, 96 eachLog ind for tilgængelighedLog ind for tilgængelighed1 * 768 Items1 * 768 Items - 440,00 kr440,00 kr
Sale Valid until 30/06/2026
Sale Valid until 30/06/20261 * 768 Items440,00 krSpecifikationer:Lav fremstillingspåvirkning More
Specifications
Cat. No.613-6036Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (Certificate Registration No. EMS 623400)Lav fremstillingspåvirkning
Racks without tips
Produktinformation
- Catalog #BeskrivelseAvailabilityPrice
- Empty rack, medium, colour code yellow and orange (for ClipTip 200 and 300)Log ind for tilgængelighed1 * 10 Items167,00 kr