Orders LinkContactUsLinkComponent
Cryogenic vials, CryoELITE®, WHEATON®
Cryogenic vials, CryoELITE®, WHEATON®
Leverandør:  DWK Life Sciences
Certifikater
Cryogenic vials, CryoELITE®, WHEATON®
Cryogenic vials, CryoELITE®, WHEATON®
30 tilgængelige muligheder nedenfor
Product Details & Documents
CryoELITE® cryogenic vials are made from low binding, cryogenic grade virgin PP. They offer an unrivaled cap seal for the shipping and storage of precious biosamples. The patented cap seal exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handling.
  • Loctagon™ Vial Skirt provides stability in freestanding position
  • Screw cap can be easily removed with one hand
  • Coloured caps allow for colour coding
  • Round bottom vials can be centrifuged up to 17 000×g
Optional 2D data matrix bar code inserts provide a unique identifier for traceability. The bottom format allows for an unrestricted view of the 2D bar code, convenient for automated scanning.
 : Lot certified RNase / DNase and Endotoxin Free providing assurance of product integrity.
Product Family Options
Freestanding, internal thread
Produktinformation
  • Kapacitet
    Grundlag
    Diameter×H
    Kasket inkluderet
    Lågfarve
    Steril
    Tilgængelighed
    Price
  • 479-0978
    With writing area
    With writing area
    1,20 ml
    Conical bottom, freestanding
    12×40 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.010,00 kr
    3.010,00 kr
    1 * 500 Items
       3.010,00 kr
  • 479-0899
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×50 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0900
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×50 mm
    Yes
    hvid
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0901
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×50 mm
    Yes
    rød
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0902
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×50 mm
    Yes
    lyserød
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0903
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×50 mm
    Yes
    gul
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0904
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×50 mm
    Yes
    grøn
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0905
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×50 mm
    Yes
    blå
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0979
    With writing area
    With writing area
    3,00 ml
    Conical bottom, freestanding
    12×63 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.200,00 kr
    4.200,00 kr
    1 * 500 Items
       4.200,00 kr
  • 479-0980
    With writing area
    With writing area
    4,00 ml
    Conical bottom, freestanding
    12×77 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.490,00 kr
    4.490,00 kr
    1 * 500 Items
       4.490,00 kr
  • 479-0981
    With writing area
    With writing area
    5,00 ml
    Conical bottom, freestanding
    12×91 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.710,00 kr
    4.710,00 kr
    1 * 500 Items
       4.710,00 kr
Freestanding, external thread
Produktinformation
  • Kapacitet
    Grundlag
    Diameter×H
    Kasket inkluderet
    Lågfarve
    Steril
    Tilgængelighed
    Price
  • 479-2073
    With writing area
    With writing area
    0,50 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    hvid
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.600,00 kr
    3.600,00 kr
    1 * 500 Items
       3.600,00 kr
  • 479-0974
    With writing area
    With writing area
    1,20 ml
    Conical bottom, freestanding
    12×37 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.010,00 kr
    3.010,00 kr
    1 * 500 Items
       3.010,00 kr
  • 479-0869
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0868
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    hvid
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0870
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    rød
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0871
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    lyserød
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0872
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    gul
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0873
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    grøn
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0874
    With writing area
    With writing area
    2,00 ml
    Conical bottom, freestanding
    12×49 mm
    Yes
    blå
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.560,00 kr
    3.560,00 kr
    1 * 500 Items
       3.560,00 kr
  • 479-0975
    With writing area
    With writing area
    3,00 ml
    Conical bottom, freestanding
    12×63 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.280,00 kr
    4.280,00 kr
    1 * 500 Items
       4.280,00 kr
  • 479-0976
    With writing area
    With writing area
    4,00 ml
    Conical bottom, freestanding
    12×78 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.580,00 kr
    4.580,00 kr
    1 * 500 Items
       4.580,00 kr
  • 479-0977
    With writing area
    With writing area
    5,00 ml
    Conical bottom, freestanding
    12×93 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.790,00 kr
    4.790,00 kr
    1 * 500 Items
       4.790,00 kr
Round bottom, internal thread
Produktinformation
  • Kapacitet
    Grundlag
    Diameter×H
    Kasket inkluderet
    Lågfarve
    Steril
    Tilgængelighed
    Price
  • 479-1139
    With writing area
    With writing area
    1,20 ml
    Round bottom
    12×39 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 2.840,00 kr
    2.840,00 kr
    1 * 500 Items
       2.840,00 kr
  • 479-1140
    With writing area
    With writing area
    2,00 ml
    Round bottom
    12×49 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.270,00 kr
    3.270,00 kr
    1 * 500 Items
       3.270,00 kr
  • 479-1143
    With writing area
    With writing area
    3,00 ml
    Round bottom
    12×63 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.200,00 kr
    4.200,00 kr
    1 * 500 Items
       4.200,00 kr
  • 479-1144
    With writing area
    With writing area
    4,00 ml
    Round bottom
    12×76 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.490,00 kr
    4.490,00 kr
    1 * 500 Items
       4.490,00 kr
  • 479-1145
    With writing area
    With writing area
    5,00 ml
    Round bottom
    12×90 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.710,00 kr
    4.710,00 kr
    1 * 500 Items
       4.710,00 kr
Round bottom, external thread
Produktinformation
  • Kapacitet
    Grundlag
    Diameter×H
    Kasket inkluderet
    Lågfarve
    Steril
    Tilgængelighed
    Price
  • 479-1135
    With writing area
    With writing area
    1,20 ml
    Round bottom
    12×35 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 2.840,00 kr
    2.840,00 kr
    1 * 500 Items
       2.840,00 kr
  • 479-1136
    With writing area
    With writing area
    2,00 ml
    Round bottom
    12×49 mm
    Yes
    Colour-21390331|Colour-21390353
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 3.270,00 kr
    3.270,00 kr
    1 * 500 Items
       3.270,00 kr
2D data matrix bar code bottom inserts
Produktinformation
  • Katalog #
    Beskrivelse
    Tilgængelighed
    Price
  • 2D data matrix bar code bottom inserts, non sterile
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 Items
    1 * 500 Items - 4.660,00 kr
    4.660,00 kr
    1 * 500 Items
       4.660,00 kr