Opsamlingsplader
Opsamlingsplade, PP, 96-brønds, V-formet bund, V-bund, 500 µl, 96
Catalog # 28-4039-43
Supplier: Cytiva
Jump to:
Product Details & Documents
Made from biocompatible PP. Collection plates, 96-well, designed for collection of eluted samples in parallel from screening plates such as MultiTrap™ and PreDictor™ plates.
- Convenient collection of samples in 96-well format
- Excellent fit to prepacked MultiTrap plates and PreDictor plates
- V-bottom minimises the loss of sample
Well volume: < 500 µl
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Form:V-bund
- Ant. brønde:96
- Volumen:500 µl
- Beskrivelse:Opsamlingsplade, PP, 96-brønds, V-formet bund