Orders LinkContactUsLinkComponent
Conductivity cells, ScienceLine
Leverandør:   SI Analytics
Conductivity cells, ScienceLine
Leverandør:  SI Analytics
Certifikater
Conductivity cells, ScienceLine
Conductivity cells, ScienceLine
2 tilgængelige muligheder nedenfor
Product Details & Documents
These high-end conductivity cells are ideal for the monitoring and control of plants as well as the examination of ultrapure water in quality control labs.
  • Models with SMEK plug head can be connected to diverse instrument types
  • Glass platinum electrodes LF713T (4-pole) and LF913T (5-pole) allow the determination of conductivity in organic media
Product Family Options
Produktinformation
  • Model
    Konduktivitet
    Cellekonstant
    Temperatur
    Materiale
    Ø×L
    Tilslutning
    Tilgængelighed
    Price
  • 663-4006
    4 pols konduktivitetscelle, sensor, 4 platinringe, temp.sensor NTC 30 kOhm
    4 pols konduktivitetscelle, sensor, 4 platinringe, temp.sensor NTC 30 kOhm
    LF 613 T
    1 µS/cm - 2000 mS/cm
    1,0 cm-1
    −5...+80 °C
    Plast
    12×120 mm
    Fast 1 m kabel med 8 pols stik
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 7.250,00 kr
    7.250,00 kr
    1 * 1 Items
       7.250,00 kr
  • 663-0282
    4 pols konduktivitetscelle, sensor, 4 platinringe, temp.sensor NTC 30 kOhm
    4 pols konduktivitetscelle, sensor, 4 platinringe, temp.sensor NTC 30 kOhm
    LF 713 T
    1 µS/cm - 2000 mS/cm
    1,0 cm-1
    −30...+135 °C
    Glas
    12×120 mm
    Fast 1 m kabel med 8 pols stik
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 8.700,00 kr
    8.700,00 kr
    1 * 1 Items
       8.700,00 kr