Double spatulas, flexible
210 mm, 11 mm
Katalog # USBE3234
Leverandør: USBECK Laborgeräte
Gå til:
Product Details & Documents
Flexible double spatulas made of stainless steel.
Specifikationer
- Længde:210 mm
- Bladbredde:11 mm
Flexible double spatulas made of stainless steel.