Double spatulas, micro
150 mm, 3 mm
Katalog # USBE3202
Leverandør: USBECK Laborgeräte
Gå til:
Product Details & Documents
Flexible micro double spatulas made of stainless steel.
Specifikationer
- Længde:150 mm
- Bladbredde:3 mm
Flexible micro double spatulas made of stainless steel.