Blotting papers
Om denne vare
Their extremely smooth surface and relatively high thickness provide them with superior absorption properties and uniformity across the entire contact area.
- Grades BF 2 and BF 3 are made of cotton linters with more than 98% alpha-cellulose content
- Four grades of blotting papers are available
Grades BF 1 and BF 4 are made of refined pulp and cotton linters with more than 95% alpha-cellulose content.
8 tilgængelige muligheder nedenfor
Product Details & Documents
Their extremely smooth surface and relatively high thickness provide them with superior absorption properties and uniformity across the entire contact area.
- Grades BF 2 and BF 3 are made of cotton linters with more than 98% alpha-cellulose content
- Four grades of blotting papers are available
Grades BF 1 and BF 4 are made of refined pulp and cotton linters with more than 95% alpha-cellulose content.
Specifications for Product Family
Specifikationer
- Catalog No:
- SARTFT-2519200200N
- 588-3851
- 588-3852
- SARTFT2-520200200N
- SARTFT2520-460570K
- 515-1606
- 515-1632
- SARTFT-3-521-185
- Vægt:
- 195 g/m²
- 195 g/m²
- 195 g/m²
- 330 g/m²
- 330 g/m²
- 330 g/m²
- 550 g/m²
- 550 g/m²
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Tykkelse:
- 0,35 mm
- 0,35 mm
- 0,35 mm
- 0,76 mm
- 0,76 mm
- 0,76 mm
- 1,3 mm
- 1,3 mm
- Miljøcertificering:
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
Specifikationer
Product Family Options
Kvalitet BF 2
Produktinformation
- DimensionerTilgængelighedPrice
- SARTFT-2519200200NPladerSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.SARTFT-2519200200NVægt195 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse0,35 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning - 588-3851PladerSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.588-3851Vægt195 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse0,35 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning - 588-3852PladerSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.588-3852Vægt195 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse0,35 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning
Kvalitet BF 3
Produktinformation
- DimensionerTilgængelighedPrice
- SARTFT2-520200200NSheetsSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.SARTFT2-520200200NVægt330 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse0,76 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning - SARTFT2520-460570KSheetsSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.SARTFT2520-460570KVægt330 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse0,76 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning - 515-1606PladerSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.515-1606Vægt330 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse0,76 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning
Kvalitet BF 4
Produktinformation
- DimensionerTilgængelighedPrice
- 515-1632PladerSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.515-1632Vægt550 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse1,3 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning - SARTFT-3-521-185DiscsSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.SARTFT-3-521-185Vægt550 g/m²Environmentally PreferableLav fremstillingspåvirkningTykkelse1,3 mmMiljøcertificeringRenewable Energy CertificateLav fremstillingspåvirkning