Hellendahl farveskåle, DURAN®
Farveskåle iht. Hellendahl, til op til 16 objektglas, 60×60×100 mm
Katalog # SCOT233150002
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Soda-Lime glass.
For up to 16 microscope slides (76x26 mm), with widening towards the top.
Advarsel: Note: Do not clean staining dishes and jars at temperatures above 60°C (glass corrosion is possible).
Specifikationer
- Beskrivelse:Farveskåle iht. Hellendahl, til op til 16 objektglas
- B×D×H:60×60×100 mm