Volumetric flasks, class A, with screw cap
Leverandør: KARTELL
Gå til:
Om denne vare
PFA, semi-transparent, med PFA skruelåg.
- High chemical resistance
- High temperature resistance (−250 to +270 °C)
- Individually calibrated ringmark
Certifications: According to DIN EN ISO 1042
4 tilgængelige muligheder nedenfor
Product Details & Documents
PFA, semi-transparent, med PFA skruelåg.
- High chemical resistance
- High temperature resistance (−250 to +270 °C)
- Individually calibrated ringmark
Certifications: According to DIN EN ISO 1042
Product Family Options
Produktinformation
- KapacitetGevindHøjdeTilgængelighedPrice