Microplates, 1536-well
HTS-mikroplade, 1536-brønds plade med faste glasindsatse, kemisk inaktiv, 50 µl
391-1972
Microtitration plate, made from PP, with fixed glass inserts.
- High purity and chemically inert
- 1536 wells
- Translucent plastic cover included in standard scope of delivery
Plate dimensions: 124×81 mm
- Beskrivelse:HTS-mikroplade, 1536-brønds plade med faste glasindsatse, kemisk inaktiv
- Brøndvolumen:50 µl