Sample vials for COULTER COUNTER®
20,00 ml, 32×56 mm, transparent, Yes
Catalog # BRND722055
Supplier: Brand
Jump to:
Product Details & Documents
Transparent PS vials with PE lid.
Specifications
- Kasket inkluderet:Yes
- Diameter×H:32×56 mm
- Kapacitet:20,00 ml
- Farve:transparent