Orders LinkContactUsLinkComponent
Flydespærring, oliesorbenter
 :   3M
Flydespærring, oliesorbenter
 :  3M
Flydespærring, oliesorbenter
Flydespærring, oliesorbenter
Booms are a selective sorbent which will absorb hydrocarbon-based liquids (oleophilic) and repel aqueous-based liquids (hydrophobic). They are used to stop the spread of liquids and are the first line in spill containment.
  • Cover large spills or as a precaution in areas where large spills may occur
  • For absorbing oil from areas where water may also be present
  • Booms can be linked together by built-in connectors to deploy over large areas
  • UV-resistant, they will not deteriorate or disintegrate
  • Cost-effective as oil, not water, is recovered
  • Can be used outside as well as in-plant
  • Mini-booms are ideal for encircling spills and containing leaks because of their tube-like shape
Product Family Options
  • Beskrivelse
    Sugeevne
    Pakket
    Ø×L
  • Flydespærring
    260 l
    4/case
    200×3000 mm
    1 * 1 Items
    1 * 1 Items - 1.270,00 kr
    1.270,00 kr
    1 * 1 Items
       1.270,00 kr
  • Dobbelt flydespærring
    152 l
    4/case
    (2×100)×3000 mm
    1 * 1 Items
    1 * 1 Items - 1.210,00 kr
    1.210,00 kr
    1 * 1 Items
       1.210,00 kr
  • Mini-flydespærring
    45 l
    12/case
    75×1200 mm
    1 * 12 Items
    1 * 12 Items - 1.890,00 kr
    1.890,00 kr
    1 * 12 Items
       1.890,00 kr