Stopcocks for burette, straight
1,5 mm, 12,5 NS
Katalog # LENZ02700501
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
NS stopcock made of borosilicate glass 3.3, facetted tip with plug of PTFE and screwthread retaining device.
- Excellent chemical resistance
- Excellent temperature resistance
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Hul Ø:1,5 mm
- Hane:12,5 NS