Orders LinkContactUsLinkComponent
Trykfiltreringsenheder, Whatman™
Trykfiltreringsenheder, Whatman™
Trykfiltreringsenheder, Whatman™
Trykfiltreringsenheder, Whatman™
Trykfiltreringsenheder, Whatman™
Pressure filtration devices with a sample loading cylinder are suitable for batch filtration of samples from 20 ml. Devices without infusion cylinders are connected inline and are suitable for larger volumes of several liters.
Filtration of liquids and gases is possible, including sterile filtration of serums or clear filtration of media that are difficult to filter, especially those that are highly viscous. Membranes, paper or glass fiber filter discs can be used. Cleaning and changing of filters is completed in a few steps.
All units are equipped with pressure-resistant filter supports. High-quality silicone or PTFE O-rings seal the systems. Please ensure that you use only intact seals for safety reasons. PTFE versions are available in addition to stainless steel devices, for use with corrosive media.
  • Catalog No:
  • 514-0352
  • WHAT10453007
  • WHAT10450450
  • WHAT10452006
  • Filtre:
  • 50 mm
  • -
  • Præfilter:
  • 43 mm
  • -
  • Max. temperature:
  • 200 °C
Product Family Options
Indløb/udløb-tilslutninger til GMP trykfiltreringsenheder af rustfrit stål i MD 050- og MD 142/5-serien
  • Beskrivelse
    Model
  • indløb/udløbs-tilslutning, MD 050/0/12, rustfrit stål, rapid seal-kobling til SV 003 b og SV 003 c
    1 * 1 Items
    1 * 1 Items - 993,00 kr
    993,00 kr
    1 * 1 Items
       993,00 kr
    Specifications
    Cat. No.
    514-0352
    Filtre
    Præfilter
    Max. temperature
  • indløb/udløbs-tilslutning, MD 050/0/18, rustfrit stål, glasøje, ekstern Ø 9 - 11 mm, til trykslanger
    1 * 1 Items
    1 * 1 Items - 1.170,00 kr
    1.170,00 kr
    1 * 1 Items
       1.170,00 kr
    Specifications
    Cat. No.
    WHAT10453007
    Filtre
    Præfilter
    Max. temperature
  • Beskrivelse
    Model
  • MD 050/4, rustfrit stål, 200 ml, 230×70 mm med rapid seal, 50 mm filter Ø
    MD 050/4
    1 * 1 Items
    1 * 1 Items - 16.800,00 kr
    16.800,00 kr
    1 * 1 Items
       16.800,00 kr
    Specifications
    Cat. No.
    WHAT10450450
    Filtre
    50 mm
    Præfilter
    43 mm
    Max. temperature
    200 °C
  • Pressure filtration device, silicon O-ring, 134×5 mm
    MD 142/0/66
    1 * 1 Items
    1 * 1 Items - 150,00 kr
    150,00 kr
    1 * 1 Items
       150,00 kr
    Specifications
    Cat. No.
    WHAT10452006
    Filtre
    -
    Præfilter
    -
    Max. temperature