Orders LinkContactUsLinkComponent
Burettes, pellet pattern, without intermediate stopcock, SILBERBRAND
 :   Brand
Burettes, pellet pattern, without intermediate stopcock, SILBERBRAND
 :  Brand
Burettes, pellet pattern, without intermediate stopcock, SILBERBRAND
Burettes, pellet pattern, without intermediate stopcock, SILBERBRAND
Amber borosilicate glass 5.4, class B.
  • Calibrated to deliver 'TD, Ex'
  • Short marks at major graduations, for 25 ml reduced distance between subdivision marks
  • Automatic zeroing, intermediate stopcock for recirculating residual liquid
25 ml: Error limits according to class B, DIN EN ISO 385
50 ml: DIN EN ISO 385, class B
*Total height includes height of reservoir bottle.
Product Family Options
Burette with titration stopcock with PTFE key without reservoir bottle
  • Kapacitet
    Inddeling
    Samlet højde
    Tolerance
  • 25 ml
    0,1 ml
    ~700* mm
    0,08 ± ml
    1 * 1 Items
    1 * 1 Items - 2.280,00 kr
    2.280,00 kr
    1 * 1 Items
       2.280,00 kr
  • 50 ml
    0,1 ml
    ~1000* mm
    0,10 ± ml
    1 * 1 Items
    1 * 1 Items - 2.280,00 kr
    2.280,00 kr
    1 * 1 Items
       2.280,00 kr
Accessories
  •  
    Beskrivelse
    Kapacitet
  • 612-9955
    Reservoir bottle, amber glass
    2000 ml
    1 * 1 Items - 994,00 kr
  •  
    Beskrivelse
    Kapacitet
  • 612-9953
    Ball set with net, black, size 5, natural rubber/PVC
    102
    1 * 1 Items - 898,00 kr
  •  
    Beskrivelse
  • BRND23400
    Enkelte gummibælge, orangefarvede, med ventil og slange
    1 * 1 Items - 156,00 kr