Flowindikator, vingehjul
SAN, 6 - 11 mm, Flow indicator paddle wheel
Catalog # BRND134080
Supplier: Brand
Jump to:
Product Details & Documents
Transparent styrene acrylonitrile (SAN) useful device to optimise cooling water flow during distillation and reduce water consumption.
Specifications
- Beskrivelse:Flow indicator paddle wheel
- Materiale:SAN
- Rør Ø:6 - 11 mm
- Cat. No.:BRND134080
- Maks. tryk:2 bar