Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, Nonane

Supelco® Organic Reference Standard, Nonane

Nonane, Analytical standard, 5 mL
Katalog # SUPL74250-5ML
Leverandør:   Merck
Supelco® Organic Reference Standard, Nonane
Katalog # SUPL74250-5ML
Leverandør:  Merck
CAS nummer:  
Supelco® Organic Reference Standard, Nonane

Nonane, Analytical standard, 5 mL

CAS nummer:  
Price
454,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents
  • GHS signal: GHS02, GHS08, GHS07, GHS09

Advarsel: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331

Specifikationer

Specifikationer

  • Pakn:
    5 mL
  • Standard:
    Analytical standard
  • Beskrivelse:
    Nonane
  • Densitet:
    0,718 g/ml at 25 °C (lit.)
  • MDL No.:
    MFCD00009574
  • Synonym(s):
    n-Nonane
  • Formel:
    CH₃(CH₂)₇CH₃
  • Cat. No.:
    SUPL74250-5ML
  • M-Clarity quality level:
    MQ100
  • CAS-nr:
    111-84-2
  • Molekylvægt:
    128,26
  • Kogepunkt:
    151 °C (lit.)
  • Fareklassebeskrivelse:
    H226 - H304 - H315 - H336 - H410
  • Flammepunkt:
    31,0 °C - Closed cup
  • Opbevaring betingelser:
    2...30 °C
  • Melting point:
    –53 °C (lit.)
  • Shipping conditions:
    STD

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.