Orders LinkContactUsLinkComponent
Membrane filters with grid
Membrane filters with grid
Membrane filters with grid
 :  Sartorius
Membrane filters with grid
Membrane filters with grid
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.
Documents
  • Catalog No:
  • 515-0187
  • SART13507-47-ACN
  • 512-5093
  • 611-0683
  • 515-0173
  • 611-1157
  • SART13106-50-ACN
  • SART13107-47-HCN
  • SART13106-47-N
  • SART13106-47-HCN
  • 515-0175
  • 512-5060
  • 512-5091
  • 512-5092
  • Model:
  • 131
  • 135
  • 131
  • 131
  • 135
  • 131
  • 131
  • 131
  • 131
  • 131
  • 135
  • 131
  • 131
  • 131
  • Tykkelse:
  • 115 - 145 µm
  • 120 µm
  • 115 - 145 µm
  • 115 - 145 µm
  • 120 µm
  • 115 - 145 µm
  • 115 - 145 µm
  • 115 - 145 µm
  • 115 - 145 µm
  • 115 - 145 µm
  • 120 µm
  • 115 - 145 µm
  • 115 - 145 µm
  • 115 - 145 µm
  • Max. temperature:
  • 130 °C
  • 180 °C
  • 130 °C
  • 130 °C
  • 180 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 180 °C
  • 130 °C
  • 130 °C
  • 130 °C
Product Family Options
Sterile, individually packaged
  • Beskrivelse
    Model
    Porestørrelse
    Ø
  • White with black grid, cellulose nitrate, 3 mm hydrophobic edge
    131
    0,20 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 2.050,00 kr
    2.050,00 kr
    1 * 100 Items
       2.050,00 kr
    Specifications
    Cat. No.
    515-0187
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • White with black grid, cellulose acetate, 3 mm hydrophobic edge
    135
    0,20 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 1.940,00 kr
    1.940,00 kr
    1 * 100 Items
       1.940,00 kr
    Specifications
    Cat. No.
    SART13507-47-ACN
    Model
    135
    Tykkelse
    120 µm
    Max. temperature
    180 °C
  • White with black grid, cellulose nitrate, 3 mm hydrophobic edge
    131
    0,20 µm
    50 mm
    1 * 100 Items
    1 * 100 Items - 2.900,00 kr
    2.900,00 kr
    1 * 100 Items
       2.900,00 kr
    Specifications
    Cat. No.
    512-5093
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • Hvid, med sort gitter
    131
    0,45 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 2.050,00 kr
    2.050,00 kr
    1 * 100 Items
       2.050,00 kr
    Specifications
    Cat. No.
    611-0683
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • Hvid, med sort gitter
    135
    0,45 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 1.580,00 kr
    1.580,00 kr
    1 * 100 Items
       1.580,00 kr
    Specifications
    Cat. No.
    515-0173
    Model
    135
    Tykkelse
    120 µm
    Max. temperature
    180 °C
  • White with black grid, cellulose nitrate, 6 mm hydrophobic edge
    131
    0,45 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 2.690,00 kr
    2.690,00 kr
    1 * 100 Items
       2.690,00 kr
    Specifications
    Cat. No.
    611-1157
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • White with black grid, cellulose nitrate, 3 mm hydrophobic edge
    131
    0,45 µm
    50 mm
    1 * 100 Items
    1 * 100 Items - 3.840,00 kr
    3.840,00 kr
    1 * 100 Items
       3.840,00 kr
    Specifications
    Cat. No.
    SART13106-50-ACN
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
Non sterile
  • Beskrivelse
    Model
    Porestørrelse
    Ø
  • White with black grid, cellulose nitrate, 6 mm hydrophobic edge
    131
    0,20 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 1.970,00 kr
    1.970,00 kr
    1 * 100 Items
       1.970,00 kr
    Specifications
    Cat. No.
    SART13107-47-HCN
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • White with black grid, cellulose nitrate, 3 mm hydrophobic edge
    131
    0,45 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 2.200,00 kr
    2.200,00 kr
    1 * 100 Items
       2.200,00 kr
    Specifications
    Cat. No.
    SART13106-47-N
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • White with black grid, cellulose nitrate, 6 mm hydrophobic edge
    131
    0,45 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 1.990,00 kr
    1.990,00 kr
    1 * 100 Items
       1.990,00 kr
    Specifications
    Cat. No.
    SART13106-47-HCN
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • Hvid, med sort gitter
    135
    0,45 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 1.910,00 kr
    1.910,00 kr
    1 * 100 Items
       1.910,00 kr
    Specifications
    Cat. No.
    515-0175
    Model
    135
    Tykkelse
    120 µm
    Max. temperature
    180 °C
  • Hvid, med sort gitter
    131
    0,45 µm
    50 mm
    1 * 100 Items
    1 * 100 Items - 1.830,00 kr
    1.830,00 kr
    1 * 100 Items
       1.830,00 kr
    Specifications
    Cat. No.
    512-5060
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • Hvid, med sort gitter
    131
    8,00 µm
    47 mm
    1 * 100 Items
    1 * 100 Items - 2.050,00 kr
    2.050,00 kr
    1 * 100 Items
       2.050,00 kr
    Specifications
    Cat. No.
    512-5091
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C
  • White with black grid, cellulose nitrate, 3 mm hydrophobic edge
    131
    8,00 µm
    50 mm
    1 * 100 Items
    1 * 100 Items - 1.940,00 kr
    1.940,00 kr
    1 * 100 Items
       1.940,00 kr
    Specifications
    Cat. No.
    512-5092
    Model
    131
    Tykkelse
    115 - 145 µm
    Max. temperature
    130 °C