Orders LinkContactUsLinkComponent
Eddikesyre
Eddikesyre
Flammable
Corrosive
Marketsource
Supplier:   Roth Carl
Eddikesyre
Supplier:  Roth Carl
Marketsource
Certificates
Eddikesyre
5 Options Available Below
Product Details & Documents
Synonyms:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS Number:64-19-7
  • Formula:H₃CCOOH
  • MDL Number:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Boiling Pt:118 °C (1013 hPa)
  • Melting Pt:17 °C
  • Density:1,05 g/cm³ (20 °C)
  • Flash Pt:38,5 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-580-7
  • UN:2789
  • ADR:8,II
  • REACH:01-2119475328-30
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L
    Specifications:
    Marketsource
    Marketsource
  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L
    Specifications:
    Marketsource
    Marketsource