Caprinsyre, Supelco®
Glass bottle for solids, 5 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Decylic acid, Decanoic acid
- CAS nummer:334-48-5
- Formel:C₁₀H₂₀O₂
- MDL nummer:MFCD00004441
- Molecular Weight:172.27 g/mol
- Merck Index:13,01764
- Kogende Pt:268 °C (1013 hPa)
- Smeltende Pt:29…32 °C
- Tæthed:0,893 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:147 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:206-376-4
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



