Drænstativ
Drænstativ med indbygget kar, 450×630 mm
BRND95105
: Brand
PS, with integral draining trough.
- Pegs can be inserted as required
- Peg holes are closed at the back
- Suitable for glass graduated cylinders of up to 250 ml
Delivery information: Supplied complete wth 72 pegs, drain hose and accessories for wall mounting.
- Beskrivelse:Drænstativ med indbygget kar
- B×H:450×630 mm