Orders LinkContactUsLinkComponent
Sprøjte/indsatsfiltre, Nalgene®
Sprøjte/indsatsfiltre, Nalgene®
   Bæredygtige muligheder
Leverandør:   Thermo Fisher Scientific
Sprøjte/indsatsfiltre, Nalgene®
   Bæredygtige muligheder
Certifikater
Sprøjte/indsatsfiltre, Nalgene®
2 tilgængelige muligheder nedenfor
Product Details & Documents
These syringe filters with PTFE membrane and PP housing are used for benchtop fermenters, in-line gas filtration for sterile air or CO₂ and to protect vacuum pumps from damage by aqueous solutions. They are also designed for filtration of aggressive chemicals and solvents such as those used in GC and HPLC.
Inlet/outlet: Stepped hose barb accepts six to nine mm Int.Ø. tubing.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 513-2221
  • 513-2222
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifikationer
Product Family Options
Produktinformation
  • Ø
    Porestørrelse
    Volumen
    Membran
    Pakket
    Tilgængelighed
    Price
  • 50 mm
    0,20 µm
    200 - 5000 ml
    PTFE
    10 pr. æske
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 1.320,00 kr
    1.320,00 kr
    1 * 10 Items
       1.320,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    513-2221
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning
  • 50 mm
    0,45 µm
    200 - 5000 ml
    PTFE
    10 pr. æske
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 1.350,00 kr
    1.350,00 kr
    1 * 10 Items
       1.350,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    513-2222
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    NAR Certificate of Retirement