UV-mikroplader, Nunc™
96-brønds plade, UV-transparent bund, klar, 342 μl, 96
Catalog # 735-0253
Supplier: Thermo Fisher Scientific
UV-mikroplader, Nunc™
Catalog # 735-0253
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Perform DNA and protein quantitation in 260 nm and 280 nm range with Thermo Scientific™ Nunc™ 96-Well UV microplates. The 96-Well format allows for a higher throughput of sample screening compared to cuvettes.
- Non sterile
- Flat bottom (F-well)
Documents
NUNC8404_TDS_EN
Datasheet
Specifications
- Ant. brønde:96
- Farve:klar
- Beskrivelse:96-brønds plade, UV-transparent bund
- Brøndvolumen:342 μl
- Cat. No.:735-0253
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning