Prednison
Glass bottle for solids, 250 mg
Katalog # PROOMM0482.00
Leverandør: LGC Standards PROMOCHEM
Gå til:
Product Details & Documents
- CAS nummer:53-03-2
- Formel:C₂₁H₂₆O₅
- MDL nummer:MFCD00003608
- Molecular Weight:358.43 g/mol
- Smeltende Pt:230…232 °C
- EINECS:200-160-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 mg



